Assay Details
Gene Symbol: PFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105813.4 | NP_565017.1 | 8 - 9 | 1099 | 79 | |
GenBank mRNA | AK118835.1 | - | 8 - 9 | 1052 | 79 | |
BT002403.1 | - | 8 - 9 | 1054 | 79 | ||
BT008888.1 | - | 8 - 9 | 1041 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
