Assay Details
| Gene Symbol: AGL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105825.3 | NP_565022.1 | 2 - 3 | 480 | 68 | |
| GenBank mRNA | AK228190.1 | - | 2 - 3 | 480 | 68 | |
| BT006157.1 | - | 2 - 3 | 478 | 68 | ||
| BT008332.1 | - | 2 - 3 | 292 | 68 | ||
| BT008524.1 | - | 2 - 3 | 292 | 68 | ||
More Information
Set Membership: |
|

AGL12
Genomic Map