Assay Details
Gene Symbol: CAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105927.4 | NP_177412.1 | 4 - 5 | 871 | 95 | |
GenBank mRNA | AY288079.1 | - | 4 - 5 | 723 | 95 | |
BT002440.1 | - | 4 - 5 | 823 | 95 | ||
BT008840.1 | - | 4 - 5 | 723 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
