Assay Details
| Gene Symbol: NF-YA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036195.5 | NP_001031272.1 | 2 - 3 | 727 | 62 | |
| NM_001334553.1 | NP_001319368.1 | 2 - 3 | 604 | 62 | ||
| NM_105941.8 | NP_565049.1 | 2 - 3 | 980 | 62 | ||
| GenBank mRNA | AY140028.1 | - | 2 - 3 | 497 | 62 | |
| BT008825.1 | - | 1 - 2 | 303 | 62 | ||
More Information
Set Membership: |
|

NF-YA3
Genomic Map