Assay Details
| Gene Symbol: NF-YA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334553.1 | NP_001319368.1 | 4 - 5 | 764 | 77 | |
| NM_105941.8 | NP_565049.1 | 4 - 5 | 1140 | 77 | ||
| GenBank mRNA | AY140028.1 | - | 4 - 5 | 657 | 77 | |
| BT008825.1 | - | 3 - 4 | 463 | 77 | ||
More Information
Set Membership: |
|

NF-YA3
Genomic Map