Assay Details
Gene Symbol: BGAL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036196.2 | NP_001031273.1 | 4 - 5 | 897 | 87 | |
NM_001334559.1 | NP_001321344.1 | 4 - 5 | 892 | 87 | ||
NM_001334560.1 | NP_001321343.1 | 5 - 6 | 749 | 87 | ||
NM_105957.3 | NP_565051.1 | 4 - 5 | 884 | 87 | ||
GenBank mRNA | AY058198.1 | - | 4 - 5 | 786 | 87 | |
AY142008.1 | - | 4 - 5 | 565 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
