Assay Details
Gene Symbol: BGAL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036196.2 | NP_001031273.1 | 7 - 8 | 1146 | 98 | |
NM_001334559.1 | NP_001321344.1 | 7 - 8 | 1141 | 98 | ||
NM_001334560.1 | NP_001321343.1 | 8 - 9 | 998 | 98 | ||
NM_105957.3 | NP_565051.1 | 7 - 8 | 1133 | 98 | ||
GenBank mRNA | AY058198.1 | - | 7 - 8 | 1035 | 98 | |
AY142008.1 | - | 7 - 8 | 814 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
