Assay Details
| Gene Symbol: PEN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344127.1 | NP_001318680.1 | 7 - 8 | 990 | 141 | |
| NM_123006.4 | NP_198464.4 | 6 - 7 | 924 | 141 | ||
More Information
Set Membership: |
|

PEN3
Genomic Map