Assay Details
| Gene Symbol: PEN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344127.1 | NP_001318680.1 | 10 - 11 | 1548 | 88 | |
| NM_123006.4 | NP_198464.4 | 9 - 10 | 1482 | 88 | ||
More Information
Set Membership: |
|

PEN3
Genomic Map