Assay Details
| Gene Symbol: AT5G36160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123007.6 | NP_198465.3 | 2 - 3 | 710 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G36160| Gene Symbol: AT5G36160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123007.6 | NP_198465.3 | 2 - 3 | 710 | 86 | |
Set Membership: |
|