Assay Details
| Gene Symbol: CIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344169.1 | NP_001331332.1 | 6 - 7 | 2649 | 231 | |
| NM_123078.3 | NP_568546.1 | 7 - 8 | 2612 | 151 | ||
More Information
Set Membership: |
|

CIP4
Genomic Map