Assay Details
| Gene Symbol: NIP4;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344221.1 | NP_001330234.1 | 3 - 4 | 819 | 53 | |
| NM_123141.2 | NP_198598.1 | 3 - 4 | 623 | 53 | ||
More Information
Set Membership: |
|

NIP4;2
Genomic Map