Assay Details
| Gene Symbol: SOS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085208.2 | NP_001078677.1 | 11 - 12 | 957 | 70 | |
| NM_001203503.1 | NP_001190432.1 | 10 - 11 | 766 | 70 | ||
| NM_001344222.1 | NP_001331291.1 | 10 - 11 | 1049 | 70 | ||
| NM_123144.4 | NP_198601.2 | 11 - 12 | 924 | 70 | ||
| GenBank mRNA | AF404865.2 | - | 10 - 11 | 728 | 70 | |
| AY084419.1 | - | 10 - 11 | 758 | 70 | ||
| AY136333.1 | - | 11 - 12 | 816 | 70 | ||
| BT008815.1 | - | 10 - 11 | 728 | 70 | ||
More Information
Set Membership: |
|

SOS4
Genomic Map