Assay Details
| Gene Symbol: SOS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085208.2 | NP_001078677.1 | 12 - 13 | 1019 | 63 | |
| NM_001203503.1 | NP_001190432.1 | 11 - 12 | 828 | 63 | ||
| NM_001344222.1 | NP_001331291.1 | 11 - 12 | 1111 | 63 | ||
| NM_123144.4 | NP_198601.2 | 12 - 13 | 986 | 63 | ||
| GenBank mRNA | AF404865.2 | - | 11 - 12 | 790 | 63 | |
| AY084419.1 | - | 11 - 12 | 820 | 63 | ||
| AY136333.1 | - | 12 - 13 | 878 | 63 | ||
| BT008815.1 | - | 11 - 12 | 790 | 63 | ||
More Information
Set Membership: |
|

SOS4
Genomic Map