Assay Details
Gene Symbol: SOS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085208.2 | NP_001078677.1 | 3 - 4 | 403 | 145 | |
NM_001203503.1 | NP_001190432.1 | 3 - 4 | 296 | 145 | ||
NM_001344222.1 | NP_001331291.1 | 2 - 3 | 495 | 145 | ||
NM_123144.4 | NP_198601.2 | 3 - 4 | 370 | 145 | ||
GenBank mRNA | AF404865.2 | - | 2 - 3 | 174 | 145 | |
AY084419.1 | - | 2 - 3 | 204 | 145 | ||
AY136333.1 | - | 3 - 4 | 262 | 145 | ||
BT008815.1 | - | 2 - 3 | 174 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
