Assay Details
| Gene Symbol: ASF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123171.4 | NP_198627.1 | 1 - 2 | 397 | 101 | |
| GenBank mRNA | AB078340.1 | - | 1 - 2 | 262 | 101 | |
| AY072420.1 | - | 1 - 2 | 281 | 101 | ||
| AY086448.1 | - | 1 - 2 | 283 | 101 | ||
| AY114716.1 | - | 1 - 2 | 112 | 101 | ||
More Information
Set Membership: |
|

ASF1B
Genomic Map