Assay Details
Gene Symbol: SYP131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337487.1 | NP_001319461.1 | 4 - 5 | 393 | 68 | |
NM_001337488.1 | NP_001326903.1 | 3 - 4 | 298 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
