Assay Details
| Gene Symbol: ATMIN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203077.1 | NP_001190006.1 | 15 - 16 | 1571 | 74 | |
| NM_001339097.1 | NP_001327198.1 | 15 - 16 | 1719 | 74 | ||
| NM_114198.4 | NP_189916.4 | 15 - 16 | 1529 | 74 | ||
More Information
Set Membership: |
|

ATMIN7
Genomic Map