Assay Details
| Gene Symbol: JAZ11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114212.5 | NP_189930.1 | 2 - 3 | 310 | 69 | |
| GenBank mRNA | BT015154.1 | - | 2 - 3 | 200 | 69 | |
| BT015661.1 | - | 2 - 3 | 186 | 69 | ||
More Information
Set Membership: |
|

JAZ11
Genomic Map