Assay Details
| Gene Symbol: JAZ11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203078.1 | NP_001190007.1 | 3 - 4 | 401 | 92 | |
| NM_114212.5 | NP_189930.1 | 6 - 7 | 709 | 92 | ||
| GenBank mRNA | BT015154.1 | - | 6 - 7 | 599 | 92 | |
| BT015661.1 | - | 6 - 7 | 585 | 92 | ||
More Information
Set Membership: |
|

JAZ11
Genomic Map