Assay Details
| Gene Symbol: HDA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114336.4 | NP_190054.2 | 2 - 3 | 407 | 62 | |
| GenBank mRNA | BT002003.1 | - | 2 - 3 | 317 | 62 | |
| BT006576.1 | - | 2 - 3 | 150 | 62 | ||
More Information
Set Membership: |
|

HDA9
Genomic Map