Assay Details
| Gene Symbol: LOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339198.1 | NP_001326504.1 | 4 - 5 | 1736 | 111 | |
| NM_114383.3 | NP_566875.1 | 4 - 5 | 1552 | 111 | ||
| GenBank mRNA | L23968.1 | - | 4 - 5 | 1385 | 111 | |
More Information
Set Membership: |
|

LOX2
Genomic Map