Assay Details
| Gene Symbol: LOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339198.1 | NP_001326504.1 | 5 - 6 | 2043 | 99 | |
| NM_114383.3 | NP_566875.1 | 5 - 6 | 1859 | 99 | ||
| GenBank mRNA | L23968.1 | - | 5 - 6 | 1692 | 99 | |
More Information
Set Membership: |
|

LOX2
Genomic Map