Assay Details
Gene Symbol: SYP72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339202.1 | NP_001319688.1 | 4 - 5 | 576 | 107 | |
NM_001339203.1 | NP_001326893.1 | 4 - 5 | 757 | 107 | ||
NM_114397.2 | NP_566878.1 | 4 - 5 | 757 | 107 | ||
GenBank mRNA | AF355758.1 | - | 4 - 5 | 576 | 107 | |
DQ446736.1 | - | 4 - 5 | 540 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
