Assay Details
| Gene Symbol: SYP72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339202.1 | NP_001319688.1 | 5 - 6 | 638 | 141 | |
| NM_001339203.1 | NP_001326893.1 | 5 - 6 | 819 | 141 | ||
| NM_114397.2 | NP_566878.1 | 5 - 6 | 819 | 141 | ||
| GenBank mRNA | AF355758.1 | - | 5 - 6 | 638 | 141 | |
| DQ446736.1 | - | 5 - 6 | 602 | 141 | ||
More Information
Set Membership: |
|

SYP72
Genomic Map