Assay Details
Gene Symbol: MLO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114398.2 | NP_566879.1 | 12 - 13 | 1330 | 91 | |
GenBank mRNA | AF369564.1 | - | 12 - 13 | 1184 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
