Assay Details
| Gene Symbol: AIR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126453.5 | NP_565309.2 | 7 - 8 | 1340 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AIR3| Gene Symbol: AIR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126453.5 | NP_565309.2 | 7 - 8 | 1340 | 105 | |
Set Membership: |
|