Assay Details
| Gene Symbol: ORG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126615.5 | NP_565333.1 | 2 - 3 | 426 | 90 | |
| GenBank mRNA | AY072356.1 | - | 2 - 3 | 311 | 90 | |
| AY114726.1 | - | 2 - 3 | 287 | 90 | ||
More Information
Set Membership: |
|

ORG4
Genomic Map