Assay Details
Gene Symbol: GRF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335301.1 | NP_001325112.1 | 1 - 2 | 362 | 85 | |
NM_126630.2 | NP_027759.1 | 1 - 2 | 299 | 85 | ||
GenBank mRNA | AY060586.1 | - | 1 - 2 | 267 | 85 | |
AY125572.1 | - | 1 - 2 | 198 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
