Assay Details
| Gene Symbol: VPS2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335305.1 | NP_001324346.1 | 4 - 5 | 954 | 57 | |
| NM_126650.4 | NP_565336.1 | 4 - 5 | 783 | 57 | ||
| GenBank mRNA | AY090953.1 | - | 4 - 5 | 664 | 57 | |
More Information
Set Membership: |
|

VPS2.1
Genomic Map