Assay Details
| Gene Symbol: HEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126675.3 | NP_565338.1 | 10 - 11 | 2519 | 132 | |
| GenBank mRNA | BT005794.1 | - | 10 - 11 | 2288 | 132 | |
More Information
Set Membership: |
|

HEN2
Genomic Map