Assay Details
| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335345.1 | NP_001323939.1 | 25 - 26 | 3007 | 77 | |
| NM_001335346.1 | NP_001323940.1 | 25 - 26 | 3007 | 77 | ||
| NM_001335347.1 | NP_001323936.1 | 24 - 25 | 2924 | 77 | ||
| NM_001335348.1 | NP_001323937.1 | 24 - 25 | 2853 | 77 | ||
| NM_001335349.1 | NP_001323938.1 | 22 - 23 | 2784 | 77 | ||
| NM_126770.8 | NP_178811.7 | 24 - 25 | 2864 | 77 | ||
More Information
Set Membership: |
|

ABCC13
Genomic Map