Assay Details
| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335345.1 | NP_001323939.1 | 31 - 32 | 4005 | 70 | |
| NM_001335346.1 | NP_001323940.1 | 32 - 33 | 3757 | 70 | ||
| NM_001335347.1 | NP_001323936.1 | 31 - 32 | 3674 | 70 | ||
| NM_001335348.1 | NP_001323937.1 | 31 - 32 | 3603 | 70 | ||
| NM_001335349.1 | NP_001323938.1 | 29 - 30 | 3534 | 70 | ||
| NM_126770.8 | NP_178811.7 | 31 - 32 | 3614 | 70 | ||
More Information
Set Membership: |
|

ABCC13
Genomic Map