Assay Details
Gene Symbol: ABCC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335345.1 | NP_001323939.1 | 12 - 13 | 1263 | 119 | |
NM_001335346.1 | NP_001323940.1 | 12 - 13 | 1263 | 119 | ||
NM_001335347.1 | NP_001323936.1 | 11 - 12 | 1180 | 119 | ||
NM_001335348.1 | NP_001323937.1 | 11 - 12 | 1109 | 119 | ||
NM_001335349.1 | NP_001323938.1 | 9 - 10 | 1040 | 119 | ||
NM_126770.8 | NP_178811.7 | 11 - 12 | 1120 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
