Assay Details
| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335345.1 | NP_001323939.1 | 15 - 16 | 1543 | 68 | |
| NM_001335346.1 | NP_001323940.1 | 15 - 16 | 1543 | 68 | ||
| NM_001335347.1 | NP_001323936.1 | 14 - 15 | 1460 | 68 | ||
| NM_001335348.1 | NP_001323937.1 | 14 - 15 | 1389 | 68 | ||
| NM_001335349.1 | NP_001323938.1 | 12 - 13 | 1320 | 68 | ||
| NM_126770.8 | NP_178811.7 | 14 - 15 | 1400 | 68 | ||
More Information
Set Membership: |
|

ABCC13
Genomic Map