Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335388.1 | NP_001318216.1 | 2 - 3 | 696 | 79 | |
| NM_126925.5 | NP_178969.1 | 2 - 3 | 766 | 79 | ||
| NM_179620.2 | NP_849951.1 | 2 - 3 | 637 | 79 | ||
| GenBank mRNA | AB048945.1 | - | 2 - 3 | 547 | 79 | |
| AF063901.1 | - | 2 - 3 | 558 | 79 | ||
| AY096498.1 | - | 2 - 3 | 556 | 79 | ||
| AY114010.1 | - | 1 - 2 | 488 | 79 | ||
More Information
Set Membership: |
|

AGT
Genomic Map