Assay Details
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335388.1 | NP_001318216.1 | 4 - 5 | 1281 | 88 | |
NM_126925.5 | NP_178969.1 | 4 - 5 | 1351 | 88 | ||
NM_179620.2 | NP_849951.1 | 4 - 5 | 1222 | 88 | ||
GenBank mRNA | AB048945.1 | - | 4 - 5 | 1132 | 88 | |
AF063901.1 | - | 4 - 5 | 1143 | 88 | ||
AY096498.1 | - | 4 - 5 | 1141 | 88 | ||
AY114010.1 | - | 3 - 4 | 1073 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
