Assay Details
| Gene Symbol: CHR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335389.1 | NP_001325087.1 | 19 - 20 | 3739 | 81 | |
| NM_001335390.1 | NP_001325086.1 | 19 - 20 | 3135 | 81 | ||
| NM_126926.5 | NP_178970.3 | 19 - 20 | 3333 | 81 | ||
More Information
Set Membership: |
|

CHR5
Genomic Map