Assay Details
| Gene Symbol: CHR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335389.1 | NP_001325087.1 | 9 - 10 | 2599 | 98 | |
| NM_001335390.1 | NP_001325086.1 | 9 - 10 | 1995 | 98 | ||
| NM_126926.5 | NP_178970.3 | 9 - 10 | 2193 | 98 | ||
More Information
Set Membership: |
|

CHR5
Genomic Map