Assay Details
Gene Symbol: ABH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126934.4 | NP_565356.1 | 8 - 9 | 879 | 80 | |
GenBank mRNA | AF268377.1 | - | 8 - 9 | 879 | 80 | |
AF272891.1 | - | 8 - 9 | 746 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -




Gene Ontology Categories:
