Assay Details
| Gene Symbol: ABCG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126940.3 | NP_178984.1 | 1 | 2254 | 96 | |
| GenBank mRNA | AY039972.1 | - | 1 | 2090 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCG5| Gene Symbol: ABCG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126940.3 | NP_178984.1 | 1 | 2254 | 96 | |
| GenBank mRNA | AY039972.1 | - | 1 | 2090 | 96 | |
Set Membership: |
|