Assay Details
Gene Symbol: SERK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335399.1 | NP_001318220.1 | 4 - 5 | 398 | 83 | |
NM_001335400.1 | NP_001324219.1 | 2 - 3 | 534 | 83 | ||
NM_001335401.1 | NP_001324220.1 | 1 - 2 | 107 | 83 | ||
NM_126956.4 | NP_179000.3 | 3 - 4 | 468 | 83 | ||
GenBank mRNA | AY094412.1 | - | 3 - 4 | 284 | 83 | |
FJ708692.1 | - | 3 - 4 | 295 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001335400.1, NM_001335401.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
