Assay Details
Gene Symbol: SERK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335399.1 | NP_001318220.1 | 7 - 8 | 686 | 61 | |
NM_001335400.1 | NP_001324219.1 | 5 - 6 | 822 | 61 | ||
NM_001335401.1 | NP_001324220.1 | 4 - 5 | 395 | 61 | ||
NM_126956.4 | NP_179000.3 | 6 - 7 | 756 | 61 | ||
GenBank mRNA | AY094412.1 | - | 6 - 7 | 572 | 61 | |
BT001116.1 | - | 3 - 4 | 232 | 61 | ||
FJ708692.1 | - | 6 - 7 | 583 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
