Assay Details
| Gene Symbol: ALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126957.2 | NP_565359.1 | 5 - 6 | 419 | 152 | |
| GenBank mRNA | AY057526.1 | - | 5 - 6 | 390 | 152 | |
| AY143898.1 | - | 5 - 6 | 347 | 152 | ||
| AY518702.1 | - | 5 - 6 | 347 | 152 | ||
More Information
Set Membership: |
|

ALD1
Genomic Map