Assay Details
| Gene Symbol: AT1G03103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202020.1 | NP_973749.1 | 1 - 2 | 340 | 75 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G03103| Gene Symbol: AT1G03103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202020.1 | NP_973749.1 | 1 - 2 | 340 | 75 | |
Set Membership: | 
		   
					
						
							 |