Assay Details
Gene Symbol: GSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100519.3 | NP_172127.1 | 11 - 12 | 1254 | 89 | |
NM_202042.3 | NP_973771.1 | 11 - 12 | 1209 | 89 | ||
GenBank mRNA | AF019927.1 | - | 11 - 12 | 1165 | 89 | |
AY035048.1 | - | 11 - 12 | 1183 | 89 | ||
AY051053.1 | - | 11 - 12 | 1140 | 89 | ||
X99696.1 | - | 1212 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
