Assay Details
Gene Symbol: GSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100519.3 | NP_172127.1 | 5 - 6 | 718 | 72 | |
NM_202042.3 | NP_973771.1 | 5 - 6 | 673 | 72 | ||
GenBank mRNA | AF019927.1 | - | 5 - 6 | 629 | 72 | |
AY035048.1 | - | 5 - 6 | 647 | 72 | ||
AY051053.1 | - | 5 - 6 | 604 | 72 | ||
X99696.1 | - | 676 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
