Assay Details
| Gene Symbol: APX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035914.2 | NP_001030991.2 | 3 - 4 | 363 | 76 | |
| NM_001084012.1 | NP_001077481.1 | 2 - 3 | 355 | 76 | ||
| NM_001084013.1 | NP_001077482.1 | 3 - 4 | 271 | 76 | ||
| NM_001123772.2 | NP_001117244.1 | 2 - 3 | 431 | 76 | ||
| NM_001331739.1 | NP_001318949.1 | 2 - 3 | 519 | 76 | ||
| NM_100663.4 | NP_172267.1 | 2 - 3 | 404 | 76 | ||
| NM_179276.2 | NP_849607.1 | 3 - 4 | 401 | 76 | ||
| NM_202057.2 | NP_973786.1 | 2 - 3 | 156 | 76 | ||
| GenBank mRNA | AK230096.1 | - | 809 | 76 | ||
| AY039879.1 | - | 2 - 3 | 207 | 76 | ||
| AY056395.1 | - | 2 - 3 | 207 | 76 | ||
| AY086425.1 | - | 2 - 3 | 260 | 76 | ||
| AY094002.1 | - | 1 - 2 | 117 | 76 | ||
| X59600.1 | - | 156 | 76 | |||
More Information
Set Membership: |
|

APX1
Genomic Map