Assay Details
| Gene Symbol: APX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035914.2 | NP_001030991.2 | 7 - 8 | 821 | 109 | |
| NM_001084012.1 | NP_001077481.1 | 6 - 7 | 813 | 109 | ||
| NM_001084013.1 | NP_001077482.1 | 7 - 8 | 729 | 109 | ||
| NM_001123772.2 | NP_001117244.1 | 6 - 7 | 889 | 109 | ||
| NM_001331739.1 | NP_001318949.1 | 6 - 7 | 977 | 109 | ||
| NM_100663.4 | NP_172267.1 | 6 - 7 | 862 | 109 | ||
| NM_179276.2 | NP_849607.1 | 7 - 8 | 859 | 109 | ||
| NM_202057.2 | NP_973786.1 | 6 - 7 | 614 | 109 | ||
| GenBank mRNA | AK230096.1 | - | 1267 | 109 | ||
| AY039879.1 | - | 6 - 7 | 665 | 109 | ||
| AY056395.1 | - | 6 - 7 | 665 | 109 | ||
| AY086425.1 | - | 6 - 7 | 718 | 109 | ||
| AY094002.1 | - | 5 - 6 | 575 | 109 | ||
| X59600.1 | - | 614 | 109 | |||
More Information
Set Membership: |
|

APX1
Genomic Map