Assay Details
| Gene Symbol: APX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035914.2 | NP_001030991.2 | 9 - 10 | 987 | 59 | |
| NM_001084013.1 | NP_001077482.1 | 9 - 10 | 895 | 59 | ||
| NM_001123772.2 | NP_001117244.1 | 8 - 9 | 1055 | 59 | ||
| NM_001331739.1 | NP_001318949.1 | 8 - 9 | 1143 | 59 | ||
| NM_100663.4 | NP_172267.1 | 8 - 9 | 1028 | 59 | ||
| NM_179276.2 | NP_849607.1 | 9 - 10 | 1025 | 59 | ||
| NM_202057.2 | NP_973786.1 | 8 - 9 | 780 | 59 | ||
| GenBank mRNA | AK230096.1 | - | 1433 | 59 | ||
| AY039879.1 | - | 8 - 9 | 831 | 59 | ||
| AY056395.1 | - | 8 - 9 | 831 | 59 | ||
| X59600.1 | - | 780 | 59 | |||
More Information
Set Membership: |
|

APX1
Genomic Map